2-Bromo-6-methoxy-3-nitropyridine 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD212877-250MGEA 6 EUR
BLDPBD212877-250MG
2-Bromo-6-methoxy-3-nitropyridine 95%
2-Bromo-6-methoxy-3-nitropyridine
Formel: C₆H₅BrN₂O₃
Molekulargewicht: 233,02 g/mol
MDL Number: MFCD07774106
CAS-Nummer: 344296-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR