Ethyl 5-bromothiophene-3-carboxylate 95%

BLDPBD212963-250MGEA 25 EUR
BLDPBD212963-250MG BLDPBD212963-5G BLDPBD212963-10G BLDPBD212963-1G
Ethyl 5-bromothiophene-3-carboxylate 95%
Ethyl 5-bromothiophene-3-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₇BrO₂S
Molekulargewicht: 235,098 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD14155650
CAS-Nummer: 170355-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR