2-Chloro-4-ethoxy-pyrimidine 95%

BLDPBD213330-250MGEA 167 EUR
BLDPBD213330-250MG BLDPBD213330-1G BLDPBD213330-100MG
2-Chloro-4-ethoxy-pyrimidine 95%
2-Chloro-4-ethoxy-pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₇ClN₂O
Molekulargewicht: 158,586 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00234199
CAS-Nummer: 83774-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR