N-Boc-DL-valin 97%

BLDPBD21393-25GEA 18 EUR
BLDPBD21393-0.025G BLDPBD21393-25G BLDPBD21393-100G BLDPBD21393-500G BLDPBD21393-0.5G BLDPBD21393-10G BLDPBD21393-5G
N-Boc-DL-valin 97%
N-Boc-DL-valin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₁₉NO₄
Molekulargewicht: 217,26 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00069973
CAS-Nummer: 54895-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR