5'-Brom-2'-methylacetanilid 98%
|
|
BLDPBD215216-5GEA
117
EUR
BLDPBD215216-5G
BLDPBD215216-1G
BLDPBD215216-25G
5'-Brom-2'-methylacetanilid 98%
5'-Brom-2'-methylacetanilid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₁₀BrNO Molekulargewicht: 228,0900 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11520466 CAS-Nummer: 116436-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...