3-Chlor-2-hydroxyphenylboronsäure 98%

BLDPBD216827-250MGEA 64 EUR
BLDPBD216827-250MG BLDPBD216827-1G BLDPBD216827-100MG
3-Chlor-2-hydroxyphenylboronsäure 98%
3-Chlor-2-hydroxyphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₆BClO₃
Molekulargewicht: 172,3740 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08689551
CAS-Nummer: 951655-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR