7-Bromo-6-methoxy-5-azaindole 97%

BLDPBD217088-100MGEA 71 EUR
BLDPBD217088-100MG BLDPBD217088-250MG BLDPBD217088-5G BLDPBD217088-1G
7-Bromo-6-methoxy-5-azaindole 97%
7-Bromo-6-methoxy-5-azaindole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇BrN₂O
Molekulargewicht: 227,058 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12962570
CAS-Nummer: 1190314-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR