Imidazo[1,2-a]pyridin-5-carbonsäure Hydrochlorid 98%

BLDPBD217397-100MGEA 34 EUR
BLDPBD217397-100MG BLDPBD217397-250MG
Imidazo[1,2-a]pyridin-5-carbonsäure Hydrochlorid 98%
Imidazo[1,2-a]pyridin-5-carbonsäure Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇ClN₂O₂
Molekulargewicht: 198,6100 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09954931
CAS-Nummer: 1198283-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR