2-Isopropoxy-3-pyridinboronsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD217888-250MGEA 9 EUR
BLDPBD217888-250MG BLDPBD217888-1G BLDPBD217888-100MG
2-Isopropoxy-3-pyridinboronsäure 95%
2-Isopropoxy-3-pyridinboronsäure

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₁₂BNO₃
Molekulargewicht: 181,0000 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD11855840
CAS-Nummer: 1150114-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR