2-Methyl-4-cyanophenylboronic acid 95%

BLDPBD218069-1GEA 84 EUR
BLDPBD218069-1G BLDPBD218069-250MG BLDPBD218069-5G
2-Methyl-4-cyanophenylboronic acid 95%
2-Methyl-4-cyanophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₈BNO₂
Molekulargewicht: 160,966 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09750473
CAS-Nummer: 313546-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR