4-Fluor-3-(isopropylcarbamoyl)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD218076-250MGEA 23 EUR
BLDPBD218076-250MG BLDPBD218076-1G BLDPBD218076-5G
4-Fluor-3-(isopropylcarbamoyl)phenylboronsäure 98%
4-Fluor-3-(isopropylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₃BFNO₃
Molekulargewicht: 225,03 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08235040
CAS-Nummer: 874219-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR