Methyl-2-amino-3-fluorbenzoat 95%

BLDPBD218466-250MGEA 18 EUR
BLDPBD218466-250MG BLDPBD218466-1G BLDPBD218466-5G BLDPBD218466-25G BLDPBD218466-10G
Methyl-2-amino-3-fluorbenzoat 95%
Methyl-2-amino-3-fluorbenzoat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₈FNO₂
Molekulargewicht: 169,1530 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11870179
CAS-Nummer: 144851-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR