6-Methoxybenzo[b]thiophene-2-carboxaldehyde 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD219144-1GEA 231 EUR
BLDPBD219144-1G BLDPBD219144-100MG BLDPBD219144-250MG
6-Methoxybenzo[b]thiophene-2-carboxaldehyde 98%
6-Methoxybenzo[b]thiophene-2-carboxaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₈O₂S
Molekulargewicht: 192,234 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11101038
CAS-Nummer: 1001203-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR