4-Chlor-3-(trifluormethyl)benzylalkohol 97%

BLDPBD22052-25GEA 105 EUR
BLDPBD22052-25G BLDPBD22052-250MG BLDPBD22052-1G BLDPBD22052-100G BLDPBD22052-10G BLDPBD22052-5G
4-Chlor-3-(trifluormethyl)benzylalkohol 97%
4-Chlor-3-(trifluormethyl)benzylalkohol

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆ClF₃O
Molekulargewicht: 210,5809 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01631538
CAS-Nummer: 65735-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR