5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester 97%

BLDPBD221323-1GEA 26 EUR
BLDPBD221323-1G BLDPBD221323-250MG BLDPBD221323-100G BLDPBD221323-5G BLDPBD221323-25G BLDPBD221323-10G
5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester 97%
5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₈N₂O₃
Molekulargewicht: 156,139 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09263987
CAS-Nummer: 85230-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR