(S)-1,2-Dimethylpiperazine 95%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD223004-1GEA
134
EUR
BLDPBD223004-1G
BLDPBD223004-5G
BLDPBD223004-250MG
BLDPBD223004-100MG
(S)-1,2-Dimethylpiperazine 95%
(S)-1,2-Dimethylpiperazine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₆H₁₄N₂ Molekulargewicht: 114,189 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11858472 CAS-Nummer: 485841-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...