(R)-α-(2-Oxiranyl)anisol 98%

BLDPBD22321-1GEA 22 EUR
BLDPBD22321-1G BLDPBD22321-25G BLDPBD22321-5G
(R)-α-(2-Oxiranyl)anisol 98%
(R)-α-(2-Oxiranyl)anisol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀O₂
Molekulargewicht: 150,1745 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06659025
CAS-Nummer: 71031-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR