4-Amino-2-hydroxypyridine 97%

BLDPBD223343-250MGEA 8 EUR
BLDPBD223343-250MG BLDPBD223343-10G BLDPBD223343-1G BLDPBD223343-100G BLDPBD223343-25G BLDPBD223343-5G
4-Amino-2-hydroxypyridine 97%
4-Amino-2-hydroxypyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₆N₂O
Molekulargewicht: 110,12 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09832889
CAS-Nummer: 38767-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR