(R)-2-Aminopropanamide hydrochloride 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD22364-25GEA 40 EUR
BLDPBD22364-25G BLDPBD22364-500G BLDPBD22364-10G BLDPBD22364-5G BLDPBD22364-1G BLDPBD22364-100G
(R)-2-Aminopropanamide hydrochloride 97%
(R)-2-Aminopropanamide hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃H₉ClN₂O
Molekulargewicht: 124,57 g/mol
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 71810-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR