5,7-Dichlor-2-methyl-8-chinolinol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD22400-25GEA 58 EUR
BLDPBD22400-25G BLDPBD22400-5G BLDPBD22400-10G BLDPBD22400-1G BLDPBD22400-100G
5,7-Dichlor-2-methyl-8-chinolinol 98%
5,7-Dichlor-2-methyl-8-chinolinol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₇Cl₂NO
Molekulargewicht: 228,0747 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00023984
CAS-Nummer: 72-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR