7-Isochinolinboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD224215-1GEA 176 EUR
BLDPBD224215-1G BLDPBD224215-250MG BLDPBD224215-5G BLDPBD224215-100MG
7-Isochinolinboronsäure 98%
7-Isoquinolineboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈BNO₂
Molekulargewicht: 172,9763 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08234617
CAS-Nummer: 1092790-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR