2-(2-Pyrrolidinyl)pyridin 98%

BLDPBD22592-250MGEA 63 EUR
BLDPBD22592-250MG BLDPBD22592-100MG BLDPBD22592-1G
2-(2-Pyrrolidinyl)pyridin 98%
2-(2-Pyrrolidinyl)pyridin

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₁₂N₂
Molekulargewicht: 148,21 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01691538
CAS-Nummer: 77790-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR