2-Cyanopyridine-5-boronic acid 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD226680-25GEA 1142 EUR
BLDPBD226680-25G BLDPBD226680-5G BLDPBD226680-250MG BLDPBD226680-1G BLDPBD226680-10G BLDPBD226680-100MG
2-Cyanopyridine-5-boronic acid 97%
2-Cyanopyridine-5-boronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₅BN₂O₂
Molekulargewicht: 147,927 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10696633
CAS-Nummer: 1011722-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR