3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester 98%

BLDPBD227391-5GEA 400 EUR
BLDPBD227391-5G BLDPBD227391-250MG BLDPBD227391-1G
3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester 98%
3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₄BrNO₃
Molekulargewicht: 312,159 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09953304
CAS-Nummer: 174184-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR