2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

BLDPBD227826-250MGEA 65 EUR
BLDPBD227826-250MG BLDPBD227826-100MG BLDPBD227826-1G
2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₈BClO₂
Molekulargewicht: 252,545 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD10698516
CAS-Nummer: 517920-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR