5-(4-Fluorophenyl)-1H-pyrazol-3-amine 95%
|
|
BLDPBD228361-1GEA
30
EUR
BLDPBD228361-1G
BLDPBD228361-25G
BLDPBD228361-5G
5-(4-Fluorophenyl)-1H-pyrazol-3-amine 95%
5-(4-Fluorophenyl)-1H-pyrazol-3-amine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₉H₈FN₃ Molekulargewicht: 177,18 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD10565813 CAS-Nummer: 72411-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...