2-(Methylsulphonyl)-10H-phenothiazine 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD228517-250MGEA 45 EUR
BLDPBD228517-250MG
2-(Methylsulphonyl)-10H-phenothiazine 98%
2-(Methylsulphonyl)-10H-phenothiazine
Formel: C₁₃H₁₁NO₂S₂
Molekulargewicht: 277,362 g/mol
MDL Number: MFCD20926232
CAS-Nummer: 23503-68-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR