5-Chlor-2-hydroxybenzophenon 98%
|
|
BLDPBD22855-5GEA
48
EUR
BLDPBD22855-5G
BLDPBD22855-250MG
BLDPBD22855-100G
BLDPBD22855-25G
BLDPBD22855-1G
5-Chlor-2-hydroxybenzophenon 98%
5-Chlor-2-hydroxybenzophenon
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₃H₉ClO₂ Molekulargewicht: 232,6600 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00020134 CAS-Nummer: 85-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...