7-Fluor-2-methylchinolin-8-boronsäure 96%

BLDPBD229309-100MGEA 39 EUR
BLDPBD229309-100MG BLDPBD229309-250MG BLDPBD229309-1G
7-Fluor-2-methylchinolin-8-boronsäure 96%
7-Fluor-2-methylchinolin-8-boronsäure

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₀H₉BFNO₂
Molekulargewicht: 204,9900 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD11504838
CAS-Nummer: 1072945-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR