1-Boc-7-methoxy-3-indolboronsäurepinakolester 95%

BLDPBD230091-250MGEA 238 EUR
BLDPBD230091-250MG BLDPBD230091-100MG BLDPBD230091-1G
1-Boc-7-methoxy-3-indolboronsäurepinakolester 95%
1-Boc-7-methoxy-3-indolboronsäurepinakolester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₀H₂₈BNO₅
Molekulargewicht: 373,2500 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD14155739
CAS-Nummer: 1218790-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR