3-Chlor-5-(ethylcarbamoyl)phenylboronsäure 96%

BLDPBD230842-100MGEA 39 EUR
BLDPBD230842-100MG BLDPBD230842-250MG
3-Chlor-5-(ethylcarbamoyl)phenylboronsäure 96%
3-Chlor-5-(ethylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₁BClNO₃
Molekulargewicht: 227,4500 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800857
CAS-Nummer: 957120-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR