3-Fluor-4-(propylcarbamoyl)phenylboronsäure 98%

BLDPBD231529-5GEA 570 EUR
BLDPBD231529-5G BLDPBD231529-1G BLDPBD231529-250MG
3-Fluor-4-(propylcarbamoyl)phenylboronsäure 98%
3-Fluor-4-(propylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₃BFNO₃
Molekulargewicht: 225,0300 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09258737
CAS-Nummer: 874289-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR