3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD231627-5GEA 503 EUR
BLDPBD231627-5G BLDPBD231627-250MG BLDPBD231627-1G
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure 98%
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁BN₂O₃S
Molekulargewicht: 250,0800 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07363799
CAS-Nummer: 871333-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR