2-Bromo-3-iodobenzyl alcohol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD231695-250MGEA 25 EUR
BLDPBD231695-250MG BLDPBD231695-1G BLDPBD231695-5G
2-Bromo-3-iodobenzyl alcohol 98%
2-Bromo-3-iodobenzyl alcohol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₆BrIO
Molekulargewicht: 312,93 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11520410
CAS-Nummer: 1261644-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR