5-Chlor-2-benzoxazolinon 99%
BLDPBD23239-25GEA
9
EUR
BLDPBD23239-25G
BLDPBD23239-10G
BLDPBD23239-5G
5-Chlor-2-benzoxazolinon 99%
5-Chlor-2-benzoxazolinon
Formel:
C₇H₄ClNO₂ Molekulargewicht: 169,57 g/mol |
MDL Number:
MFCD00005717 CAS-Nummer: 95-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...