[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex 98%

BLDPBD23246-1GEA 56 EUR
BLDPBD23246-0.025G BLDPBD23246-1G BLDPBD23246-10G BLDPBD23246-25G BLDPBD23246-5G BLDPBD23246-100G
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex 98%
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₃₅H₃₀Cl₄FeP₂Pd
Molekulargewicht: 816,64 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00792899
CAS-Nummer: 95464-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR