2-(Trimethylsilylethinyl)pyridin 95%
|
|
BLDPBD233521-250MGEA
10
EUR
BLDPBD233521-250MG
BLDPBD233521-25G
BLDPBD233521-5G
BLDPBD233521-1G
2-(Trimethylsilylethinyl)pyridin 95%
2-(Trimethylsilylethinyl)pyridin
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₀H₁₃NSi Molekulargewicht: 175,31 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00066349 CAS-Nummer: 86521-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...