4-Fluorbenzoesäureanhydrid 95%

BLDPBD234452-250MGEA 16 EUR
BLDPBD234452-250MG BLDPBD234452-1G BLDPBD234452-25G BLDPBD234452-5G BLDPBD234452-10G
4-Fluorbenzoesäureanhydrid 95%
4-Fluorobenzoic anhydride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₈F₂O₃
Molekulargewicht: 262,21 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00425938
CAS-Nummer: 25569-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR