4-Isobutylphenylboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD235999-250MGEA 22 EUR
BLDPBD235999-250MG BLDPBD235999-1G
4-Isobutylphenylboronsäurepinakolester 98%
4-Isobutylphenylboronsäurepinakolester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₂₅BO₂
Molekulargewicht: 260,1795 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05663842
CAS-Nummer: 1033753-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR