4-Chloro-3-isopropoxyphenylboronic acid 95%

BLDPBD236156-1GEA 101 EUR
BLDPBD236156-1G
4-Chloro-3-isopropoxyphenylboronic acid 95%
4-Chloro-3-isopropoxyphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₂BClO₃
Molekulargewicht: 214,454 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD17015746
CAS-Nummer: 1256346-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR