5-Bromo-2,3-dimethyl-2H-indazole 95%

BLDPBD236437-1GEA 89 EUR
BLDPBD236437-1G
5-Bromo-2,3-dimethyl-2H-indazole 95%
5-Bromo-2,3-dimethyl-2H-indazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrN₂
Molekulargewicht: 225,085 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12028634
CAS-Nummer: 878064-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR