3-Bromo-5-chlorophenylboronic acid 95%
|
|
BLDPBD236756-10GEA
71
EUR
BLDPBD236756-10G
BLDPBD236756-1G
BLDPBD236756-25G
BLDPBD236756-100G
BLDPBD236756-250MG
BLDPBD236756-5G
3-Bromo-5-chlorophenylboronic acid 95%
3-Bromo-5-chlorophenylboronic acid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₆H₅BBrClO₂ Molekulargewicht: 235,271 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13189469 CAS-Nummer: 1186403-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...