4-Cyano-2-methoxybenzoic acid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD236931-5GEA 662 EUR
BLDPBD236931-5G BLDPBD236931-250MG BLDPBD236931-100MG BLDPBD236931-1G
4-Cyano-2-methoxybenzoic acid 98%
4-Cyano-2-methoxybenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇NO₃
Molekulargewicht: 177,157 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11520909
CAS-Nummer: 89469-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR