Methyl 5-chloro-6-methylpyrazine-2-carboxylate 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD237689-100MGEA 14 EUR
BLDPBD237689-100MG BLDPBD237689-1G BLDPBD237689-250MG BLDPBD237689-5G
Methyl 5-chloro-6-methylpyrazine-2-carboxylate 97%
Methyl 5-chloro-6-methylpyrazine-2-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇ClN₂O₂
Molekulargewicht: 186,596 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD18633146
CAS-Nummer: 77168-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR