4-Brom-3-methyl-1H-indazol 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD237717-25GEA 684 EUR
BLDPBD237717-25G BLDPBD237717-5G BLDPBD237717-1G BLDPBD237717-250MG
4-Brom-3-methyl-1H-indazol 95%
4-Bromo-3-methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇BrN₂
Molekulargewicht: 211,0586 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09743499
CAS-Nummer: 1159511-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR