4-Chlor-N,N-dimethylbenzamid 98%

BLDPBD238158-5GEA 129 EUR
BLDPBD238158-5G BLDPBD238158-25G BLDPBD238158-1G BLDPBD238158-10G
4-Chlor-N,N-dimethylbenzamid 98%
4-Chlor-N,N-dimethylbenzamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀ClNO
Molekulargewicht: 183,6400 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00463859
CAS-Nummer: 14062-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR