1-Brom-4-(brommethyl)naphthalin 97%

BLDPBD240306-5GEA 137 EUR
BLDPBD240306-5G BLDPBD240306-25G BLDPBD240306-10G BLDPBD240306-1G BLDPBD240306-250MG
1-Brom-4-(brommethyl)naphthalin 97%
1-Bromo-4-(bromomethyl)naphthalene

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₈Br₂
Molekulargewicht: 299,9892 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07363811
CAS-Nummer: 79996-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR