Methyl 2-Acetylpyridine-4-carboxylate 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD240786-5GEA 55 EUR
BLDPBD240786-5G BLDPBD240786-250MG BLDPBD240786-100G BLDPBD240786-1G BLDPBD240786-25G BLDPBD240786-10G
Methyl 2-Acetylpyridine-4-carboxylate 98%
Methyl 2-Acetylpyridine-4-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉NO₃
Molekulargewicht: 179,173 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09032412
CAS-Nummer: 138715-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR