1,5-Dimethyl-1H-pyrazol-4-boronsäure 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD241953-100MGEA
40
EUR
BLDPBD241953-100MG
BLDPBD241953-250MG
1,5-Dimethyl-1H-pyrazol-4-boronsäure 98%
1,5-Dimethyl-1H-pyrazole-4-boronic acid
Storage condition: Sealed in dry, Store in freezer, under -20°C
Formel:
C₅H₉BN₂O₂ Molekulargewicht: 139,9482 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD13195678 CAS-Nummer: 1204333-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...