2-Methyl-3-pyridinboronsäure Hydrochlorid 97%

BLDPBD242507-1GEA 35 EUR
BLDPBD242507-1G BLDPBD242507-250MG BLDPBD242507-5G
2-Methyl-3-pyridinboronsäure Hydrochlorid 97%
2-Methyl-3-pyridinboronsäure Hydrochlorid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₉BClNO₂
Molekulargewicht: 173,4100 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06659489
CAS-Nummer: 1072952-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR